missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-2-Benzyloxycyclohexylamine, ChiPros 99+%, ee 99%

Product Code. p-7046979
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393906 1 g 1g
11303916 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393906 Supplier Thermo Scientific Alfa Aesar Supplier No. L17020.03
Add to basket
This item is not returnable. View return policy

CAS: 216394-07-9 | C13H19NO | 205.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216394-07-9
Molecular Formula C13H19NO
Molecular Weight (g/mol) 205.30
MDL Number MFCD01075753
InChI Key NTHNRYLIXJZHRZ-UHFFFAOYNA-N
PubChem CID 11908838
IUPAC Name (1S,2S)-2-phenylmethoxycyclohexan-1-amine
SMILES NC1CCCCC1OCC1=CC=CC=C1

Specifications

Density 1.02
Boiling Point 79°C to 80°C (0.08 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5275
Quantity 1 g
UN Number UN2735
Beilstein 8981794
Sensitivity Air Sensitive
Synonym 1s,2s-2-benzyloxy cyclohexanamine, 1s,2s-+-2-benzyloxycyclohexylamine, 1s,2s-2-benzyloxycyclohexylamine, 1s,2s-2-benzyloxy cyclohexan-1-amine, 1s,2s-trans-2-benzyloxy-cyclohexylamine, 1s, 2s-2-benzyloxycyclohexylamine, 1s,2s-2-benzyloxy-cyclohexylamine, 1s, 2s-2-benzyloxy-cyclohexylamine, 1s,2s-2-benzyloxy cyclohexylamine, 1s,2s-1-amino-2-benzyloxycyclohexane
Optical Rotation +85° (Neat)
Formula Weight 205.3
Percent Purity ≥99%
Assay ee 99%
Chemical Name or Material (1S,2S)-(+)-2-Benzyloxycyclohexylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.