missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-2-Benzyloxycyclohexyl isothiocyanate, 97%

Product Code. 15480867
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15480867

Brand: Thermo Scientific Alfa Aesar L20060.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 737000-89-4 | C14H17NOS | 247.356 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 737000-89-4
Melting Point 33°C to 36°C
Density 1.106
Molecular Formula C14H17NOS
Refractive Index 1.5674
MDL Number MFCD05664047
Quantity 1 g
UN Number UN2923
Sensitivity Moisture sensitive
Synonym 1s,2s-+-2-benzyloxycyclohexyl isothiocyanate, 1s,2s-2-isothiocyanatocyclohexyl oxy methyl benzene, 1s,2s-2-phenylmethoxy cyclohexanisothiocyanate
InChI Key DNRPSBSMXLYTQF-KBPBESRZSA-N
SMILES C1CCC(C(C1)N=C=S)OCC2=CC=CC=C2
IUPAC Name [(1S,2S)-2-isothiocyanatocyclohexyl]oxymethylbenzene
Molecular Weight (g/mol) 247.356
PubChem CID 11863573
Formula Weight 247.36
Percent Purity 97%
Chemical Name or Material (1S,2S)-(+)-2-Benzyloxycyclohexyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.