missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-2-Amino-1-phenylpropyldiphenylphosphine, 97%, Thermo Scientific™

Product Code. 11925941 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass bottle
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11925941 100 mg Glass bottle 100mg
11935941 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11925941 Supplier Thermo Scientific Acros Supplier No. 434141000
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 341968-71-6
Molecular Formula C21H22NP
Molecular Weight (g/mol) 319.39
MDL Number MFCD11044864
InChI Key JWZAIGGNEGTDMG-AURADERDNA-N
Synonym 1s,2s-1-diphenylphosphino-1-phenylpropan-2-amine, 1s,2s-2-amino-1-phenylpropyl diphenylphosphine, 1s,2s-2-amino-1-phenylpropyldiphenylphosphine, 1s,2s-1-diphenylphosphanyl-1-phenylpropan-2-amine, 1s,2s-2-diphenylphosphino-1-methyl-2-phenylethylamine, 1s,2s-2-amino-1-phenylpropyl diphenylphosphane, 1s,2s-1-diphenylphosphino-1-phenyl-2-propanamine, 1s,2s-2-amino-1-phenylpropyldiphenylphosphine,min, s,s-+-2-amino-1-phenylpropyldiphenylphosphine, 1s,2s-2-amino-1-phenylpropyl diphenylphosphine, kanata purity
PubChem CID 46176707
IUPAC Name (1S,2S)-1-diphenylphosphanyl-1-phenylpropan-2-amine
SMILES C[C@H](N)[C@@H](P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 100.0°C to 105.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 mg
Formula Weight 319.38
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (1S, 2S)-2-Amino-1-phenylpropyldiphenylphosphine
Show More Show Less
compliance-icons
Product Identifier
  • (1S,2S)-2-Amino-1-phenylpropyldiphenylphosphine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.