missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-2-Amino-1-phenyl-1,3-propanediol, 97%

Product Code. 11339837
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339837 5 g 5g
11349837 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339837 Supplier Thermo Scientific Alfa Aesar Supplier No. 042183.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Asymetric synthesis of chiral 2-oxazolines | CAS: 28143-91-1 | C9H13NO2 | 167.208 g/mol

Asymetric synthesis of chiral 2-oxazolines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28143-91-1
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00004503
InChI Key JUCGVCVPNPBJIG-IUCAKERBSA-N
PubChem CID 119881
IUPAC Name (1S,2S)-2-amino-1-phenylpropane-1,3-diol
SMILES C1=CC=C(C=C1)C(C(CO)N)O

Specifications

Melting Point 112°C to 115°C
Quantity 5 g
Synonym 1s,2s-+-2-amino-1-phenyl-1,3-propanediol, 1s,2s-2-amino-1-phenylpropane-1,3-diol, l-+-threo-2-amino-1-phenyl-1,3-propanediol, 1s,2s-2-amino-1-phenyl-1,3-propanediol, 1s,2s-2-amino-1-phenyl-propane-1,3-diol, 1s, 2s-+-2-amino-1-phenyl-1,3-propanediol, 1,3-propanediol, 2-amino-1-phenyl-, 1s,2s, pubchem6415, s r*,r*-2-amino-1-phenylpropane-1,3-diol, s,s-2-amino-1-phenyl-1,3-propanediol
Formula Weight 167.21
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material (1S,2S)-(+)-2-Amino-1-phenyl-1,3-propanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.