missing translation for 'onlineSavingsMsg'
Learn More

(1S,2R)-2-(3,4-Difluorophenyl)-cyclopropanamine (S)-(+)-Mandelic Acid, TRC

Product Code. 30387586
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387586 500 mg 500mg
30381223 250 mg 250mg
30374890 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387586 Supplier Toronto Research Chemicals Supplier No. TRCD445463500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1S,2R)-2-(3,4-Difluorophenyl)-cyclopropanamine (S)-(+)-Mandelic Acid
Analyte or Component Names (1S,2R)-2-(3,4-Difluorophenyl)-cyclopropanamine (S)-(+)-Mandelic Acid
Shipping Condition Room Temperature
Molecular Formula C9H9F2N • C8H8O3
InChI Formula InChI=1S/C9H9F2N.C8H8O3/c10-7-2-1-5(3-8(7)11)6-4-9(6)12;9-7(8(10)11)6-4-2-1-3-5-6/h1-3,6,9H,4,12H2;1-5,7,9H,(H,10,11)/t6-,9+;7-/m10/s1
Solution Type Neat
SMILES FC1=C(F)C=CC([C@@H]2[C@@H](N)C2)=C1.O[C@H](C(O)=O)C3=CC=CC=C3
IUPAC Name (1S,2R)-2-(3,4-difluorophenyl)cyclopropan-1-amine (S)-2-hydroxy-2-phenylacetic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 321.32
Quantity 500 mg
Formula Weight 321.12
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.