missing translation for 'onlineSavingsMsg'
Learn More

(1S,2R)-2-(2-Benzylamino-1-hydroxyethyl)-6-fluorochromane, TRC

Product Code. 30136396
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136396 5 mg 5mg
17952765 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136396 Supplier Toronto Research Chemicals Supplier No. TRCB2259355MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129050-27-7
Molecular Formula C18H20FNO2
Molecular Weight (g/mol) 301.36
Synonym 2H-1-Benzopyran-2-methanol, 6-fluoro-3,4-dihydro-α-[[(phenylmethyl)amino]methyl]-, (αS,2R)-, 2H-1-Benzopyran-2-methanol, 6-fluoro-3,4-dihydro-α-[[(phenylmethyl)amino]methyl]-, [R-(R*,S*)]-, (αS,2R)-6-Fluoro-3,4-dihydro-α-[[(phenylmethyl)amino]methyl]-2H-1-benzopyran-2-methanol, (-)-(S)-N-Benzyl-2-[(R)-6-fluorochroman-2-yl]-2-hydroxyethanamine, (1S)-1-((2R)-6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-yl)-2-[(phenylmethyl)amino]ethanol, 2H-1-Benzopyran-2-methanol 6-fluoro-3,4-dihydro-α-[[(phenylmethyl)amino]methyl]-, (αS,2R)-, (1S,2R)-2-(2-Benzylamino-1-hydroxyethyl)-6-fluorochromane
IUPAC Name (1S)-2-(benzylamino)-1-[(2R)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]ethanol
SMILES O[C@@H](CNCc1ccccc1)[C@H]2CCc3cc(F)ccc3O2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C18H20FNO2/c19-15-7-9-17-14(10-15)6-8-18(22-17)16(21)12-20-11-13-4-2-1-3-5-13/h1-5,7,9-10,16,18,20-21H,6,8,11-12H2/t16-,18+/m0/s1
Formula Weight 301.1478
Chemical Name or Material (1S,2R)-2-(2-Benzylamino-1-hydroxyethyl)-6-fluorochromane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.