missing translation for 'onlineSavingsMsg'
Learn More

(1S,2R)-(-)-10,2-Camphorsultam, 99%

Product Code. 11432628
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packungsgröße:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
11432628 25 g 25g
11412628 1 g 1g
11422628 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11432628 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. A15897.14
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 94594-90-8 | C10H17NO2S | 215.311 g/mol

(1S,2R)-(-)-10,2-Camphorsultam is used in the asymmetric synthesis of (S)- and (R)-N-Fmoc-S-trityl-α-methylcysteine. It is used as proton source in the synthesis of chiral α,γ-substituted γ-butyrolactones. It is also employed as a chiral probe for the optical resolution by HPLC and X-ray crystallographic determination of the absolute stereochemistry of carboxylic acids. Also used to prepare N-acryloyl derivatives which are employed as dienophiles in asymmetric Diels-Alder reactions. and for other asymmetric transformations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S,2R)-(-)-10,2-Camphorsultam is used in the asymmetric synthesis of (S)- and (R)-N-Fmoc-S-trityl-α-methylcysteine. It is used as proton source in the synthesis of chiral α,γ-substituted γ-butyrolactones. It is also employed as a chiral probe for the optical resolution by HPLC and X-ray crystallographic determination of the absolute stereochemistry of carboxylic acids. Also used to prepare N-acryloyl derivatives which are employed as dienophiles in asymmetric Diels-Alder reactions. and for other asymmetric transformations.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 94594-90-8
Molecular Formula C10H17NO2S
Molecular Weight (g/mol) 215.311
MDL Number MFCD00066271
InChI Key DPJYJNYYDJOJNO-SZBHIRRCSA-N
Synonym 5r,7r-10,10-dimethyl-3??-thia-4-azatricyclo 5.2.1.0 1 ,? decane-3,3-dione, --10,2-camphorsultam;--exo-10,2-bornanesultam; 3as-3a, a,6, a,7a, a-hexahydro-8,8-dimethyl-3h-3a,6-methano-2,1-benzisothiazole-2,2-dioxide; 1s,5r-10,10-dimethyl-3-thia-4-azatricyclo 5.2.1.01,5 decane 3,3-dioxide
PubChem CID 54579826
SMILES CC1(C2CCC13CS(=O)(=O)NC3C2)C

Specifications

Melting Point 181°C to 186°C
Quantity 25 g
Beilstein 83811
Solubility Information Slightly soluble in water.
Optical Rotation −33° (c=1 in Ethanol)
Formula Weight 215.32
Percent Purity 99%
Chemical Name or Material (1S,2R)-(-)-10,2-Camphorsultam

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.