Learn More
(1S,2R)-(-)-10,2-Camphorsultam, 99%
Description
(1S,2R)-(-)-10,2-Camphorsultam is used in the asymmetric synthesis of (S)- and (R)-N-Fmoc-S-trityl-α-methylcysteine. It is used as proton source in the synthesis of chiral α,γ-substituted γ-butyrolactones. It is also employed as a chiral probe for the optical resolution by HPLC and X-ray crystallographic determination of the absolute stereochemistry of carboxylic acids. Also used to prepare N-acryloyl derivatives which are employed as dienophiles in asymmetric Diels-Alder reactions. and for other asymmetric transformations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(1S,2R)-(-)-10,2-Camphorsultam is used in the asymmetric synthesis of (S)- and (R)-N-Fmoc-S-trityl-α-methylcysteine. It is used as proton source in the synthesis of chiral α,γ-substituted γ-butyrolactones. It is also employed as a chiral probe for the optical resolution by HPLC and X-ray crystallographic determination of the absolute stereochemistry of carboxylic acids. Also used to prepare N-acryloyl derivatives which are employed as dienophiles in asymmetric Diels-Alder reactions. and for other asymmetric transformations.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
Specifications
Specifications
| Melting Point | 181°C to 186°C |
| Quantity | 1 g |
| Beilstein | 83811 |
| Solubility Information | Slightly soluble in water. |
| Optical Rotation | −33° (c=1 in Ethanol) |
| Formula Weight | 215.32 |
| Percent Purity | 99% |
| Chemical Name or Material | (1S,2R)-(-)-10,2-Camphorsultam |
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.