missing translation for 'onlineSavingsMsg'
Learn More

(1S)-[1,1'-Binaphthalene]-2,2'-diol, TRC

Product Code. 30333882
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30333882 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333882 Supplier Toronto Research Chemicals Supplier No. TRCB387020100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1S)-[1,1'-Binaphthalene]-2,2'-diol
CAS 18531-99-2
Molecular Formula C20H14O2
Quantity 100 g
Synonym [1,1'-Binaphthalene]-2,2'-diol, (S)-, (S)-(-)-[1,1'-Binaphthalene]-2,2'-diol, (-)-(1S)-[1,1'-Binaphthalene]-2,2'-diol, (-)-1,1'-Bi-2-naphthol, (-)-1,1'-Binaphthyl-2,2'-diol, (-)-1,1'-Bis(2-naphthol), (-)-2,2'-Binaphthol, (-)-2,2'-Dihydroxy-1,1'-binaphthalene, (-)-2,2'-Dihydroxy-1,1'-binaphthyl, (-)-2,2'-Dihydroxy-1,1'-dinaphthyl, (-)-Bi-beta-naphthol, (-)-Binol, (-)-S-1,1'-Binaphthyl-2,2'-diol, (1S)-1,1'-Binaphthalene-2,2'-diol, (S)-(-)-1,1'-Bi-2-naphthol, (S)-(-)-2,2'-Dihydroxy-1,1'-binaphthalene, (S)-(-)-2,2'-Dihydroxy-1,1'-binaphthyl, (S)-(-)-BINOL, (S)-(-)-Bi-2-naphthol, (S)-(-)-[1,1'-Binaphthalene]-2,2'-diol, (S)-1,1'-Bi-2,2'-naphthol, (S)-1,1'-Bi-2-naphthol, (S)-1,1'-Binaphthol, (S)-1,1'-Binaphthyl-2,2'-diol, (S)-1,1'-Bis-2-naphthol, (S)-2,2'-Dihydroxy-1,1'-binaphthalene, (S)-2,2'-Dihydroxy-1,1'-binaphthyl, (S)-BINOL, (S)-Binaphthol, (S)-[1,1'-Binaphthalene]-2,2'-diol
InChI Formula InChI=1S/C20H14O2/c21-17-11-9-13-5-1-3-7-15(13)19(17)20-16-8-4-2-6-14(16)10-12-18(20)22/h1-12,21-22H
SMILES Oc1ccc2ccccc2c1c3c(O)ccc4ccccc34
IUPAC Name 1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol
Molecular Weight (g/mol) 286.32
Formula Weight 286.0994
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.