missing translation for 'onlineSavingsMsg'
Learn More

[(1RS,4SR)-7,7-Dimethyl-2-oxobicyclo[2.2.1]hept-1-yl]methanesulfonic Acid ((+/-)-10-Camphorsulfonic Acid), Mikromol™

Product Code. 30050754
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30050754

Brand: Mikromol MM1094.070025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

[(1RS,4SR)-7,7-Dimethyl-2-oxobicyclo[2.2.1]hept-1-yl]methanesulfonic Acid ((+/-)-10-Camphorsulfonic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-10-Camphorsulfonic Acid
CAS 8-2-5872
Analyte or Component Names DL-10-Camphorsulfonic Acid
Shipping Condition Room Temperature
Molecular Formula C10 H16 O4 S
Certifications/Compliance ISO 9001
Synonym Bicyclo[2.2.1]heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, 10-Bornanesulfonic acid, 2-oxo-, (±)- (8CI), Bicyclo[2.2.1]heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, (±)-, (RS)-10-Camphorsulfonic acid, (±)-Camphor-10-sulfonic acid, (±)-Camphorsulfonic acid, 2-Oxo-10-bornanesulfonic acid, Camphor-10-sulfonic acid, DL-10-Camphorsulfonic acid, DL-Camphorsulfonic acid, Voriconazole EP impurity E, [(1RS,4SR)-7,7-Dimethyl-2-oxobicyclo[2.2.1]hept-1-yl]methanesulfonic Acid, (+/-)-10-Camphorsulfonic Acid, Ecamsule Related Compound D, Voriconazole Related Compound F, Voriconazole Impurity E
InChI Formula InChI=1S/C10H16O4S/c1-9(2)7-3-4-10(9,8(11)5-7)6-15(12,13)14/h7H,3-6H2,1-2H3,(H,12,13,14)/t7-,10-/m0/s1
SMILES CC1(C)[C@H]2CC[C@]1(CS(=O)(=O)O)C(=O)C2
IUPAC Name [(1S,4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
Molecular Weight (g/mol) 232.3
Quantity 25 mg
Formula Weight 232.0769 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.