missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1RS,2S)-Nicotine N,N'-Dioxide (3-[(1RS,2S)-1-Methyl-1-oxidopyrrolidin-2-yl]pyridine 1-Oxide), Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
(1RS,2S)-Nicotine N,N'-Dioxide (3-[(1RS,2S)-1-Methyl-1-oxidopyrrolidin-2-yl]pyridine 1-Oxide)
Specifications
Specifications
| Chemical Name or Material | (1RS,2S)-Nicotine N,N'-Dioxide |
| CAS | 2055-29-0 |
| Analyte or Component Names | (1RS,2S)-Nicotine N,N'-Dioxide |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C10 H14 N2 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Pyridine, 3-[(2S)-1-methyl-1-oxido-2-pyrrolidinyl]-, 1-oxide, Nicotine, 1,1'-dioxide (7CI,8CI), Nicotine, N,N'-dioxide (6CI), Pyridine, 3-(1-methyl-2-pyrrolidinyl)-, N,1-dioxide, (2S)-, Dioxynicotine, Pyridine, 3-(1-methyl-1-oxido-2-pyrrolidinyl)-, 1-oxide, (2S)-, S-(-)-Nicotine-1,1'-dioxide, (1RS,2S)-Nicotine N,N'-Dioxide (3-[(1RS,2S)-1-Methyl-1-oxidopyrrolidin-2-yl]pyridine 1-Oxide) |
| IUPAC Name | 3-[(2S)-1-Methyl-1-oxido-2-pyrrolidinyl]pyridine 1-oxide |
| Molecular Weight (g/mol) | 194.23 |
| Quantity | 100 mg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction