missing translation for 'onlineSavingsMsg'
Learn More

(1RS)-1-(6-Methoxy-2-naphthyl)ethanol (Naproxen Impurity K), TRC

Product Code. 17993995
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17993995 50 mg 50mg
17985005 25 mg 25mg
17974935 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17993995 Supplier Toronto Research Chemicals Supplier No. TRCM26474050MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77301-42-9
Molecular Formula C13H14O2
Molecular Weight (g/mol) 202.25
Synonym (1RS)-1-(6-Methoxynaphthalen-2-yl)ethanol, Naproxen Imp. K (EP)
IUPAC Name 1-(6-methoxynaphthalen-2-yl)ethanol
SMILES COc1ccc2cc(ccc2c1)C(C)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H14O2/c1-9(14)10-3-4-12-8-13(15-2)6-5-11(12)7-10/h3-9,14H,1-2H3
Formula Weight 202.0994
Percent Purity >95%
Chemical Name or Material (1RS)-1-(6-Methoxy-2-naphthyl)ethanol (Naproxen Impurity K)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.