missing translation for 'onlineSavingsMsg'
Learn More

(1RS)-1-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-yl)ethanol, Mikromol™

Product Code. 18020660
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18020660 100 mg 100mg
17929669 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18020660 Supplier Mikromol Supplier No. MM0281.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(1RS)-1-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-yl)ethanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(2,4-Dichlorophenyl)-2-imidazol-1-yl ethanol
CAS 24155-42-8
Analyte or Component Names 1-(2,4-Dichlorophenyl)-2-imidazol-1-yl ethanol
Shipping Condition Room Temperature
Molecular Formula C11 H10 Cl2 N2 O
Certifications/Compliance ISO 17025
Synonym Econazole Nitrate Imp. A (EP), Enilconazole Imp. E (EP), Isoconazole Nitrate Imp. A (EP), Miconazole Nitrate Imp. A (EP), Tioconazole Imp. D (EP), Econazole USP Related Compound A, Econazole USP RC A, (1RS)-1-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-yl)ethanol, Econazole Related Compound A, Econazole Nitrate Impurity A, Enilconazole Impurity E, Isoconazole Impurity B, Isoconazole Nitrate Impurity A, Miconazole Impurity A, Miconazole Nitrate Impurity A, Sertaconazole Nitrate Impurity A, Tioconazole Impurity D, Econazole Impurity A, Isoconazole Impurity A, Miconazole Impurity A, Sertaconazole Impurity A
InChI Formula InChI=1S/C11H10Cl2N2O/c12-8-1-2-9(10(13)5-8)11(16)6-15-4-3-14-7-15/h1-5,7,11,16H,6H2
SMILES OC(Cn1ccnc1)c2ccc(Cl)cc2Cl
IUPAC Name 1-(2,4-dichlorophenyl)-2-imidazol-1-ylethanol
Molecular Weight (g/mol) 257.12
Quantity 100 mg
Formula Weight 256.017 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.