missing translation for 'onlineSavingsMsg'
Learn More

1R-trans-Permethrinic Acid, TRC

Product Code. 30133631
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133631 10 g 10g
30008035 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133631 Supplier Toronto Research Chemicals Supplier No. TRCP28771510G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55701-03-6
Molecular Formula C8 H10 Cl2 O2
Molecular Weight (g/mol) 209.07
Synonym Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, (1R,3S)-, Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, (1R-trans)-, (1R)-(+)-trans-Permethrinic acid, 1R-trans-Permethric acid, 1R-trans-Permethrinic acid, Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, (1R-trans)-, (1R,3S)-3-(2,2-Dichloroethenyl)-2,2-dimethylcyclopropanecarboxylic acid, (1R)-(+)-trans-Permethrinic acid, (1R)-trans-3-(2,2-Dichloro-1-ethenyl)-2,2-dimethylcyclopropanecarboxylic acid, (1R)-trans-3-(2,2-Dichloroethenyl)-2,2-dimethylcyclopropanecarboxylic acid, 1R-trans-Permethric acid, 1R-trans-Permethrinic acid, Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, (1R,3S)-
IUPAC Name (1R,3S)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylic acid
SMILES CC1(C)[C@@H](C=C(Cl)Cl)[C@@H]1C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H10Cl2O2/c1-8(2)4(3-5(9)10)6(8)7(11)12/h3-4,6H,1-2H3,(H,11,12)/t4-,6+/m1/s1
Formula Weight 208.0058
Chemical Name or Material 1R-trans-Permethrinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.