missing translation for 'onlineSavingsMsg'
Learn More

(1R,E)-(+)-Camphorquinone 3-oxime, 99%, Thermo Scientific Chemicals

Product Code. 15497917
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15497917 5 g 5g
15487917 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497917 Supplier Thermo Scientific Alfa Aesar Supplier No. H51034.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31571-14-9
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.235
MDL Number MFCD00074848
InChI Key YRNPDSREMSMKIY-WKEGUHRASA-N
Synonym anti-1r-+-camphorquinone3-oxime, 1s,4r-2-hydroxyimino-4,7,7-trimethylbicyclo 2.2.1 heptan-3-one
PubChem CID 23308125
IUPAC Name (1R,4R)-2-hydroxyimino-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C2CCC1(C(=O)C2=NO)C)C

Specifications

Quantity 5 g
Beilstein 3200377
Formula Weight 181.23
Percent Purity 99%
Chemical Name or Material (1R,E)-(+)-Camphorquinone 3-oxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.