missing translation for 'onlineSavingsMsg'
Learn More

(1R,E)-(+)-Camphorquinone 3-oxime, 99%, Thermo Scientific Chemicals

Catalog Number 15487917
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
31571-14-9
C10H15NO2
181.235
MFCD00074848
YRNPDSREMSMKIY-WKEGUHRASA-N
anti-1r-+-camphorquinone3-oxime, 1s,4r-2-hydroxyimino-4,7,7-trimethylbicyclo 2.2.1 heptan-3-one
23308125
(1R,4R)-2-hydroxyimino-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
CC1(C2CCC1(C(=O)C2=NO)C)C
This item is not returnable. View return policy
31571-14-9
C10H15NO2
181.235
MFCD00074848
YRNPDSREMSMKIY-WKEGUHRASA-N
anti-1r-+-camphorquinone3-oxime, 1s,4r-2-hydroxyimino-4,7,7-trimethylbicyclo 2.2.1 heptan-3-one
23308125
(1R,4R)-2-hydroxyimino-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
CC1(C2CCC1(C(=O)C2=NO)C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Beilstein 3200377
Formula Weight 181.23
Percent Purity 99%
Chemical Name or Material (1R,E)-(+)-Camphorquinone 3-oxime

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'