missing translation for 'onlineSavingsMsg'
Learn More

(1R-cis)-Cyhalothric Acid, TRC

Product Code. 17913805
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17913805 100 mg 100mg
17943935 25 mg 25mg
17914915 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17913805 Supplier Toronto Research Chemicals Supplier No. TRCC422075100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76023-99-9
Molecular Formula C9 H10 Cl F3 O2
Molecular Weight (g/mol) 242.62
Synonym Bifenthrin free acid metabolite, (1R,3R)cyclopropanecarboxylic acid, -3-[(1Z)2-Chloro-3,3,3-trifluoropropen-1-yl]-2,2-dimethyl, Cyclopropanecarboxylic acid, 3-(2-chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-, [1R-[1α,3α(Z)]]-, Cyclopropanecarboxylic acid, 3-[(1Z)-2-chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethyl-, (1R,3R)- (9CI), (1R)-cis-3-[(Z)-2-Chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethylcyclopropanecarboxylic acid, (1R-cis)-Cyhalothric acid
IUPAC Name (1R,3R)-3-[(Z)-2-chloro-3,3,3-trifluoroprop-1-enyl]-2,2-dimethylcyclopropane-1-carboxylic acid
SMILES CC1(C)[C@@H](\C=C(/Cl)\C(F)(F)F)[C@H]1C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H10ClF3O2/c1-8(2)4(6(8)7(14)15)3-5(10)9(11,12)13/h3-4,6H,1-2H3,(H,14,15)/b5-3-/t4-,6-/m0/s1
Formula Weight 242.0321
Chemical Name or Material Bifenthrin free acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.