missing translation for 'onlineSavingsMsg'
Learn More

(1R)-(+)-Camphor, 98%

Product Code. 11434290
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434290 500 g 500g
11414290 25 g 25g
11424290 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434290 Supplier Thermo Scientific Alfa Aesar Supplier No. A10708.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 464-49-3 | C10H16O | 152.24 g/mol

(1R)-(+)-Camphor is used as a chiral intermediate and auxiliary. It is also used as a skin antipruritic and as an anti-infective agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R)-(+)-Camphor is used as a chiral intermediate and auxiliary. It is also used as a skin antipruritic and as an anti-infective agent.

Solubility
Soluble in water (0.1 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 464-49-3
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00064149,MFCD00074738,MFCD00064149,MFCD00064149
InChI Key DSSYKIVIOFKYAU-XVKPBYJWSA-N
Synonym +-camphor, d-camphor, 1r-+-camphor, r-camphor, +-bornan-2-one, r-+-camphor, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, 1r,4r-camphor, camphor d, +-2-bornanone
PubChem CID 159055
ChEBI CHEBI:15396
IUPAC Name (1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C)[C@H]2CC[C@@]1(C)C(=O)C2

Specifications

Melting Point 175°C to 180°C
Density 0.99
Boiling Point 204°C
Flash Point 65°C (149°F)
Odor Camphor-like
Quantity 500 g
UN Number UN2717
Beilstein 2042745
Merck Index 14,1732
Solubility Information Soluble in water (0.1g/L at 20°C).
Optical Rotation +43° (c=10 in ethanol)
Formula Weight 152.24
Percent Purity 98%
Chemical Name or Material (1R)- (+)-Camphor
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (1R)-(+)-Camphor
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: (1R)-(+)-Camphor

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.