missing translation for 'onlineSavingsMsg'
Learn More

(1R)-(-)-Camphor-10-sulfonic acid, 98%

Product Code. 11467134
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467134 25 g 25g
11477134 100 g 100g
11487134 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467134 Supplier Thermo Scientific Alfa Aesar Supplier No. B21553.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35963-20-3 | C10H16O4S | 232.29 g/mol

(1R)-(-)-Camphor-10-sulfonic acid, is used as a pharamaceutical intermediate and also used as a chiral derivative of Camp. It can also be used as resolving agents for chiral amines and other cations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R)-(-)-Camphor-10-sulfonic acid, is used as a pharamaceutical intermediate and also used as a chiral derivative of Camp. It can also be used as resolving agents for chiral amines and other cations.

Solubility
Soluble in water, and a wide variety of organic substances.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35963-20-3
Molecular Formula C10H16O4S
Molecular Weight (g/mol) 232.29
MDL Number MFCD00150753,MFCD00064158
InChI Key MIOPJNTWMNEORI-MHPPCMCBSA-N
Synonym l--camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, 1r---camphor-10-sulfonic acid, unii-y6075i4fxe, camphorsulfonic acid,-, l---camphor-10-sulfonic acid, s-camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 hept-1-yl methanesulfonic acid, --camphor-10-sulfonic acid
PubChem CID 5771688
ChEBI CHEBI:55401
SMILES CC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2

Specifications

Melting Point ∼200°C
Quantity 25 g
UN Number UN3261
Beilstein 2809676
Sensitivity Hygroscopic
Solubility Information Soluble in water,and a wide variety of organic substances.
Optical Rotation −21° (c=10 in Water)
IUPAC Name [(1R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonic acid
Formula Weight 232.3
Percent Purity ≥98%
Chemical Name or Material (1R)-(-)-Camphor-10-sulfonic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.