missing translation for 'onlineSavingsMsg'
Learn More

(1R)-(+)-alpha-Pinene, 98%, 80% ee

Product Code. p-184601 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
250 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
250g
500g
This item is not returnable. View return policy

Product Code. 10133140

Brand: Thermo Scientific Acros 131261000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7785-70-8 | C10H16 | 136.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7785-70-8
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00001346
InChI Key GRWFGVWFFZKLTI-RKDXNWHRSA-N
Synonym +-alpha-pinene, 1r,5r-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1r-+-alpha-pinene, 1r,5r-alpha-pinene, unii-h6cm4twh1w, +-pinene, h6cm4twh1w, 1r,5r-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1r,5r, +-pin-2 3-ene
PubChem CID 82227
ChEBI CHEBI:28261
SMILES CC1=CC[C@@H]2C[C@H]1C2(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -62.0°C
Color Colorless
Density 0.8600g/mL
Boiling Point 155.0°C
Flash Point 33°C
Enantiomeric Excess 75% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4650 to 1.4670
Quantity 100 g
Beilstein 05, 146
Fieser 15,201
Merck Index 14, 7445
Specific Gravity 0.86
Specific Rotation Condition +43.50° (20°C neat)
Specific Rotation + 43.50
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,chloroform,diethyl ether
IUPAC Name (1R,5R)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene
Formula Weight 136.24
Percent Purity 98%,80% ee
Physical Form Liquid
Chemical Name or Material (1R)-(+)-α-Pinene, 98% (80+% e.e.)
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (1R)-(+)-alpha-Pinene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.