missing translation for 'onlineSavingsMsg'
Learn More

(1R,3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee

Product Code. p-70147 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10222712 250 mg Glass Bottle 250mg
10374352 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10222712 Supplier Thermo Scientific Acros Supplier No. 361832500
Add to basket
This item is not returnable. View return policy

CAS: 261165-05-3 | C11H19NO4 | 229.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261165-05-3
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.27
MDL Number MFCD01320857
InChI Key RNJQBGXOSAQQDG-JGVFFNPUSA-N
Synonym 1s,3r-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, +-1s,3r-n-boc-3-aminocyclopentanecarboxylic acid, 1r,3s-n-boc-1-aminocyclopentane-3-carboxylic acid, 1s,3r-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, cis-3-boc-amino cyclopentane-1-carboxylic acid, 1s,3r-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1s,3r-3-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid
PubChem CID 1512529
IUPAC Name (1S,3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCC(C1)C(=O)O

Specifications

Melting Point 111.8°C
CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (ELSD) (HPLC)
Packaging Glass Bottle
Solubility Information Solubility in water: soluble
Quantity 250 mg
Formula Weight 229.27
Percent Purity 95%,98% ee
Physical Form Powder and/or Chunks
Chemical Name or Material (1R, 3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.