missing translation for 'onlineSavingsMsg'
Learn More

(1R,3S)-(+)-4-Cyclopentene-1,3-diol 1-acetate, 99%, stabilized

Product Code. 10603605 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10603605 5 g Glass bottle 5g
10557254 250 mg Glass bottle 250mg
10182764 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10603605 Supplier Thermo Scientific Acros Supplier No. 399340050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60410-16-4 | C7H10O3 | 142.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60410-16-4
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.15
MDL Number MFCD00078933
InChI Key IJDYOKVVRXZCFD-RQJHMYQMSA-N
Synonym 1s,4r-cis-4-acetoxy-2-cyclopenten-1-ol, 1r,4s-4-hydroxycyclopent-2-en-1-yl acetate, 1r,4s-4-hydroxycyclopent-2-enyl acetate, 1r,3s-+-cis-4-cyclopentene-1,3-diol 1-acetate, acetic acid 1r,4s-4-hydroxy-cyclopent-2-enyl ester, 1r,3s-+-4-cyclopentene-1,3-diol 1-acetate, 1s,4r-4-acetoxy-2-cyclopenten-1-ol, 1r,3s-4-cyclopentene-1,3-diol 1-acetate, 1r,4s-cis-4-hydroxy-2-cyclopentenyl acetate, 5beta-acetoxycyclopentene-3beta-ol
PubChem CID 10154103
IUPAC Name [(1R,4S)-4-hydroxycyclopent-2-en-1-yl] acetate
SMILES CC(=O)OC1CC(C=C1)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 43°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation +67° to +69° (20°C, 589nm) ((c=2.3 in chloroform))
Formula Weight 142.15
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material (1R, 3S)-(+)-4-Cyclopentene-1, 3-diol 1-acetate, stabilzed
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.