missing translation for 'onlineSavingsMsg'
Learn More

(1R,3S)-(-)-3-Aminocyclopentanecarboxylic acid, 95%, Thermo Scientific™

Product Code. 15499595
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15499595

Brand: Thermo Scientific Alfa Aesar H52122.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(1R,3S)-(-)-3-Aminocyclopentanecarboxylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71830-08-5
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00211289
InChI Key MLLSSTJTARJLHK-UHNVWZDZSA-N
Synonym 1r,3s-3-aminocyclopentanecarboxylic acid, 1r,3s-3-aminocyclopentane-1-carboxylic acid, --1r,3s-3-aminocyclopentanecarboxylic acid, cis-3-aminocyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-amino-, 1r,3s, 1r,3s---3-aminocyclopentanecarboxylic acid, cis-3-amino-cyclopentanecarboxylic acid, 1r,3s-3-amino-cyclopentanecarboxylic acid, pubchem18731, 1r,3s---3-amino-cyclopentanecarboxylic acid
PubChem CID 1502035
IUPAC Name (1R,3S)-3-aminocyclopentane-1-carboxylic acid
SMILES C1CC(CC1C(=O)O)N

Specifications

Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 129.16
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material (1R,3S)-(-)-3-Aminocyclopentanecarboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.