missing translation for 'onlineSavingsMsg'
Learn More

(1R,3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee

Product Code. 10068911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10068911 500 mg 500mg
10763983 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10068911 Supplier Thermo Scientific Acros Supplier No. 362395000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 489446-85-7 | C11H19NO4 | 229.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 489446-85-7
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.27
MDL Number MFCD02259727
InChI Key RNJQBGXOSAQQDG-HTQZYQBOSA-N
Synonym 1r,3r-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,3r-n-boc-1-aminocyclopentane-3-carboxylic acid, trans-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1s,3r-+-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3r-n-boc-3-aminocyclopentane carboxylic acid, 1r,3r-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxy carbonylamino cyclopentanecarboxylic acid
PubChem CID 1512527
IUPAC Name (1R,3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCC(C1)C(=O)O

Specifications

Melting Point 117.3°C
CAS Min % 94.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Solubility Information Solubility in water: soluble
Quantity 500 mg
Formula Weight 229.27
Percent Purity 95%,98% ee
Physical Form Powder
Chemical Name or Material (1R, 3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.