missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S)-(+)-cis-2-(Benzylamino)cyclohexanemethanol, 99%, 99% e.e., Thermo Scientific™

Product Code. 10663795
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 71581-92-5
Molecular Formula C14H21NO
Molecular Weight (g/mol) 219.33
MDL Number MFCD00151395
InChI Key BRQFIORUNWWNBM-KBPBESRZSA-N
Synonym 1r,2s-2-benzylamino cyclohexyl methanol, +-cis-2-benzylaminocyclohexanemethanol, cis-1r,2s-+-2-benzylamino cyclohexanemethanol, --cis-2-benzylaminocyclohexanemethanol, cis-1r,2s-+-2-benzylaminocyclohexanemethanol, pubchem7939, cis-2-benzylamino cyclohexylmethanol, 2-benzylamino cyclohexyl methanol #, cis-2-benzylamino cyclohexyl methanol
PubChem CID 853687
IUPAC Name [(1R,2S)-2-(benzylamino)cyclohexyl]methanol
SMILES C1CCC(C(C1)CO)NCC2=CC=CC=C2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 66°C to 69°C
Color White
Enantiomeric Excess 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2NHC6H10CH2OH
Quantity 5 g
Formula Weight 219.33
Percent Purity 99%,99% e.e
Physical Form Crystalline Powder
Chemical Name or Material (1R, 2S)-(+)-cis-2-(Benzylamino)cyclohexanemethanol, 99% e.e.
Show More Show Less
compliance-icons
Product Identifier
  • (1R,2S)-(+)-cis-2-(Benzylamino)cyclohexanemethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'