missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S)-(+)-cis-1-Amino-2-indanol, 98%

Product Code. 10078611 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10078611 10 g Glass bottle 10g
10793003 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10078611 Supplier Thermo Scientific Acros Supplier No. 297530100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 136030-00-7 | C9H12NO | 150.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136030-00-7
Molecular Formula C9H12NO
Molecular Weight (g/mol) 150.20
MDL Number MFCD00216656,MFCD01318239
InChI Key LOPKSXMQWBYUOI-DTWKUNHWSA-O
Synonym 1r,2s-1-amino-2-indanol, 1r,2s-1-amino-2,3-dihydro-1h-inden-2-ol, cis-1-amino-2-indanol, 1r,2s-+-cis-1-amino-2-indanol, cis-1-amino-2,3-dihydro-1h-inden-2-ol, 1r,2s-+-1-amino-2-hydroxyindan, 1r,2s-1-amino-2-hydroxyindane, 1r,2s-+-1-amino-2-indanol, 1r,2s-+-cis-1-aminoindan-2-ol, 1r,2s-cis-1-amino-2-indanol
PubChem CID 2725045
IUPAC Name (1R,2S)-2-hydroxy-2,3-dihydro-1H-inden-1-aminium
SMILES [NH3+][C@H]1[C@@H](O)CC2=CC=CC=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 117°C to 121°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
Specific Rotation Condition +44.5° (20°C c=1,methanol)
Specific Rotation + 44.5
Solubility Information Solubility in water: soluble
Formula Weight 149.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (1R, 2S)-(+)-cis-1-Amino-2-indanol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • (1R,2S)-(+)-cis-1-Amino-2-indanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.