missing translation for 'onlineSavingsMsg'
Learn More

(1R, 2S, 5S)-tert-Butyl Edoxaban, TRC

Product Code. 30380061
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380061 50 mg 50mg
30380739 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30380061 Supplier Toronto Research Chemicals Supplier No. TRCE48051050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1R, 2S, 5S)-tert-Butyl Edoxaban
CAS 480452-36-6
Molecular Formula C21 H30 Cl N5 O5
Quantity 50 mg
Synonym tert-Butyl ((1R,2S,5S)-2-(2-((5-chloropyridin-2-yl)amino)-2-oxoacetamido)-5-(dimethylcarbamoyl)cyclohexyl)carbamate
InChI Formula InChI=1S/C21H30ClN5O5/c1-21(2,3)32-20(31)25-15-10-12(19(30)27(4)5)6-8-14(15)24-17(28)18(29)26-16-9-7-13(22)11-23-16/h7,9,11-12,14-15H,6,8,10H2,1-5H3,(H,24,28)(H,25,31)(H,23,26,29)/t12-,14-,15+/m0/s1
SMILES CN(C)C(=O)[C@H]1CC[C@H](NC(=O)C(=O)Nc2ccc(Cl)cn2)[C@@H](C1)NC(=O)OC(C)(C)C
IUPAC Name tert-butyl N-[(1R,2S,5S)-2-[[2-[(5-chloropyridin-2-yl)amino]-2-oxoacetyl]amino]-5-(dimethylcarbamoyl)cyclohexyl]carbamate
Molecular Weight (g/mol) 467.946
Formula Weight 467.194
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.