missing translation for 'onlineSavingsMsg'
Learn More

(1R, 2S, 5S)-2-Amino Edoxaban, TRC

Product Code. 30386562
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386562 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386562 Supplier Toronto Research Chemicals Supplier No. TRCE4807905MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1R, 2S, 5S)-2-Amino Edoxaban
CAS 480450-71-3
Molecular Formula C17 H27 N5 O2 S
Quantity 5 mg
Synonym (1R, 2S, 5S)-2-Amino Edoxaban
InChI Formula InChI=1S/C17H27N5O2S/c1-21(2)17(24)10-4-5-11(18)13(8-10)19-15(23)16-20-12-6-7-22(3)9-14(12)25-16/h10-11,13H,4-9,18H2,1-3H3,(H,19,23)/t10-,11-,13+/m0/s1
SMILES CN(C)C(=O)[C@H]1CC[C@H](N)[C@@H](C1)NC(=O)c2nc3CCN(C)Cc3s2
IUPAC Name N-[(1R,2S,5S)-2-amino-5-(dimethylcarbamoyl)cyclohexyl]-5-methyl-6,7-dihydro-4H-[1,3]thiazolo[5,4-c]pyridine-2-carboxamide
Molecular Weight (g/mol) 365.494
Formula Weight 365.189
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.