missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S,5R)-(-)-Menthol, TRC

Product Code. 30133095
Change view
Click to view available options
Quantity:
1 g
5 g
250 g
Unit Size:
1g
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133095 5 g 5g
30137738 250 g 250g
17904785 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133095 Supplier Toronto Research Chemicals Supplier No. TRCM2188755G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2216-51-5
Molecular Formula C10 H20 O
Molecular Weight (g/mol) 156.27
Synonym Levomenthol, (-)-Menthol, Cyclohexanol, 5-methyl-2-(1-methylethyl)-, (1R,2S,5R)-, Cyclohexanol, 5-methyl-2-(1-methylethyl)-, [1R-(1α,2β,5α)]-, Menthol, (1R,3R,4S)-(-)- (8CI), (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexanol, (-)-(1R,3R,4S)-Menthol, (-)-Menthyl alcohol, (-)-trans-p-Menthan-cis-3-ol, (1R)-(-)-Menthol, (1R,2S,5R)-(-)-Menthol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexan-1-ol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexan-1-ol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, (1R,2S,5R)-5-Methyl-2-(propan-2-yl)cyclohexanol, (R)-(-)-Menthol, 1R-Menthol, A 10474, FEMA 2665, Hakkano, L-Menthol, L-Mentholum, Minclea, NSC 62788, l-(-)-Menthol, l-Menthol
IUPAC Name (1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol
SMILES CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10-/m1/s1
Formula Weight 156.1514
Percent Purity >95%
Chemical Name or Material (-)-Menthol
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.