missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S)-(-)-2-Amino-1,2-diphenylethanol, 99%, Thermo Scientific Chemicals

Product Code. 10264834
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23190-16-1
Molecular Formula C14H15NO
Molecular Weight (g/mol) 213.28
MDL Number MFCD00074960
InChI Key GEJJWYZZKKKSEV-UONOGXRCSA-N
Synonym 1r,2s-2-amino-1,2-diphenylethanol, 1r,2s---2-amino-1,2-diphenylethanol, 1r,2s-2-amino-1,2-diphenyl-ethanol, 1r,2s-2-amino-1,2-diphenylethan-1-ol, 1s,2r-1,2-diphenyl-2-hydroxyethylamine, pubchem5986, ksc201s9n, gejjwyzzkkksev-uonogxrcsa, erythro-2-amino-1,2-diphenylethanol, 1r,2s---diphenyl-2-aminoethanol
PubChem CID 719819
IUPAC Name (1R,2S)-2-amino-1,2-diphenylethanol
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)N

Specifications

Melting Point 142.0°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2(NH2)CH2(OH)C6H5
Quantity 1 g
Formula Weight 213.28
Percent Purity 99%
Chemical Name or Material (1R,2S)-(-)-2-Amino-1,2-diphenylethanol
compliance-icons
Product Identifier
  • (1R,2S)-(-)-2-Amino-1,2-diphenylethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'