missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(-)-N-(4-Toluenesulfonyl)-1,2-diphenylethylenediamine, 98%, Thermo Scientific™

Product Code. 10140662 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10140662 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10140662 Supplier Thermo Scientific Acros Supplier No. 363685000
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 144222-34-4
CAS Min % 97.5
CAS Max % 100.0
Melting Point 126.0°C to 130.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C21H23N2O2S
MDL Number MFCD02093428
Quantity 500 mg
Specific Rotation Condition − 61.50 (20.00°C c=1,CH2Cl2)
Specific Rotation − 61.50
Synonym 1r,2r---n-4-toluenesulfonyl-1,2-diphenylethylenediamine, 1r,2r---n-p-tosyl-1,2-diphenylethylenediamine, r,r-tsdpen, r,r-n-p-toluenesulfonyl-1,2-diphenylethylenediamine, 1r,2r-n-p-tosyl-1,2-diphenylethylenediamine, n-1r,2r-2-amino-1,2-diphenylethyl-4-methylbenzenesulfonamide, r,r-n-2-amino-1,2-diphenylethyl-p-toluenesulfonamide, 1r,2r---n-p-tosyl-1,2-diphenylethylene diamine, 1r, 2r---n-4-toluene sulfonyl 1,2-diphenyl-1,2-ethane diamine, benzenesulfonamide, n-1r,2r-2-amino-1,2-diphenylethyl-4-methyl
Solubility Information Solubility in water: insoluble.
InChI Key UOPFIWYXBIHPIP-NHCUHLMSSA-O
SMILES CC1=CC=C(C=C1)S(=O)(=O)N[C@@H]([C@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name N-[(1R,2R)-2-amino-1,2-diphenylethyl]-4-methylbenzenesulfonamide
Molecular Weight (g/mol) 367.49
PubChem CID 2734565
Formula Weight 366.48
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (1R, 2R)-(-)-N-(4-Toluenesulfonyl)-1, 2-diphenylethylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.