Learn More
(1R,2R)-Cyclohexane-1,2-dicarboxylic acid, 98+%
Description
Chiral dicarboxylic acid guests with a two C spacer (1R,2R)-cyclohexane-1,2-dicarboxylic acid and Boc-L-aspartic acid] gave CD-active species.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral dicarboxylic acid guests with a two C spacer (1R,2R)-cyclohexane-1,2-dicarboxylic acid and Boc-L-aspartic acid] gave CD-active species.
Solubility
Soluble in acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Quantity | 5 g |
| Solubility Information | Soluble in acetone. |
| Formula Weight | 172.18 |
| Percent Purity | ≥98% |
| Chemical Name or Material | (1R,2R)-Cyclohexane-1,2-dicarboxylic acid |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.