missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-Cyclohexane-1,2-dicarboxylic acid, 98+%

Product Code. 15449715
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15449715

Brand: Thermo Scientific Alfa Aesar H52293.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 46022-05-3 | C8H12O4 | 172.18 g/mol

Chiral dicarboxylic acid guests with a two C spacer (1R,2R)-cyclohexane-1,2-dicarboxylic acid and Boc-L-aspartic acid] gave CD-active species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chiral dicarboxylic acid guests with a two C spacer (1R,2R)-cyclohexane-1,2-dicarboxylic acid and Boc-L-aspartic acid] gave CD-active species.

Solubility
Soluble in acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 46022-05-3
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00062973
InChI Key QSAWQNUELGIYBC-PHDIDXHHSA-N
Synonym 1r,2r-cyclohexane-1,2-dicarboxylic acid, trans-1,2-cyclohexanedicarboxylic acid, trans-cyclohexane-1,2-dicarboxylic acid, 1r,2r---1,2-cyclohexanedicarboxylic acid, 1r,2r-1,2-cyclohexanedicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, 1r,2r-rel, 1,2-cyclohexanedicarboxylic acid, 1r,2r, 1r.2r-cyclohexane-1,2-dicarboxylicacid, 1,2-cyclohexanedicarboxylic acid, trans, pubchem19603
PubChem CID 720895
IUPAC Name (1R,2R)-cyclohexane-1,2-dicarboxylic acid
SMILES C1CCC(C(C1)C(=O)O)C(=O)O

Specifications

Quantity 250 mg
Solubility Information Soluble in acetone.
Formula Weight 172.18
Percent Purity ≥98%
Chemical Name or Material (1R,2R)-Cyclohexane-1,2-dicarboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.