missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R,5S,6R,9R,10S)-rel-1,2,5,6,9,10-Hexabromocyclododecane, TRC

Product Code. 30390162
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390162 10 mg 10mg
30398704 25 mg -
30396647 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390162 Supplier Toronto Research Chemicals Supplier No. TRCH29411010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134237-50-6
Molecular Formula C12 H18 Br6
Molecular Weight (g/mol) 641.7
Synonym alpha-Hexabromocyclododecane, alpha-1,2,5,6,9,10-Hexabromocyclododecane, Cyclododecane, 1,2,5,6,9,10-hexabromo-, (1R,2R,5S,6R,9R,10S)-rel-, Cyclododecane, 1,2,5,6,9,10-hexabromo-, (1R*,2R*,5S*,6R*,9R*,10S*)-, rel-(1R,2R,5S,6R,9R,10S)-1,2,5,6,9,10-Hexabromocyclododecane, (±)-α-HBCD, (±)-α-Hexabromocyclododecane, α-HBCD, α-Hexabromocyclododecane, α-1,2,5,6,9,10-Hexabromocyclododecane
IUPAC Name (1S,2R,5R,6S,9R,10R)-1,2,5,6,9,10-hexabromocyclododecane
SMILES Br[C@@H]1CC[C@H](Br)[C@H](Br)CC[C@@H](Br)[C@@H](Br)CC[C@H]1Br

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H18Br6/c13-7-1-2-8(14)10(16)5-6-12(18)11(17)4-3-9(7)15/h7-12H,1-6H2/t7-,8-,9+,10+,11-,12-/m1/s1
Formula Weight 635.6509
Percent Purity >95%
Chemical Name or Material alpha-Hexabromocyclododecane
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.