missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R,5R,6S,9S,10R)-rel-1,2,5,6,9,10-Hexabromocyclododecane, TRC

Product Code. 30398026
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30398026 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398026 Supplier Toronto Research Chemicals Supplier No. TRCH29410025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 134237-52-8
Molecular Formula C12 H18 Br6
Quantity 25 mg
Synonym Cyclododecane, 1,2,5,6,9,10-hexabromo-, (1R,2R,5R,6S,9S,10R)-rel-, Cyclododecane, 1,2,5,6,9,10-hexabromo-, (1R*,2R*,5R*,6S*,9S*,10R*)-, rel-(1R,2R,5R,6S,9S,10R)-1,2,5,6,9,10-Hexabromocyclododecane, (±)-γ-HBCD, (±)-γ-Hexabromocyclododecane, γ-HBCD, γ-Hexabromocyclododecane, gamma-Hexabromocyclododecane, gamma-1,2,5,6,9,10-Hexabromocyclododecane
InChI Formula InChI=1S/C12H18Br6/c13-7-1-2-8(14)10(16)5-6-12(18)11(17)4-3-9(7)15/h7-12H,1-6H2/t7-,8-,9-,10+,11-,12+/m1/s1
SMILES Br[C@@H]1CC[C@@H](Br)[C@@H](Br)CC[C@H](Br)[C@H](Br)CC[C@H]1Br
IUPAC Name (1R,2R,5R,6S,9S,10R)-1,2,5,6,9,10-hexabromocyclododecane
Molecular Weight (g/mol) 641.7
Formula Weight 635.6509
Percent Purity >95%
Chemical Name or Material gamma-Hexabromocyclododecane
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.