missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R,3S,5R)-(-)-2,3-Pinanediol, 99%, Thermo Scientific Chemicals

Catalog Number 10349250
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
22422-34-0
C10H18O2
170.25
MFCD09955216
MOILFCKRQFQVFS-UHFFFAOYNA-N
1r,2r,3s,5r---2,3-pinanediol, 1r,2r,3s,5r---pinanediol, 1r,2r,3s,5r-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, --2,3-pinanediol, unii-r58l0w3a75, --cis-pinane-cis-2,3-diol, pinanediol, --2-hydroxyisopinocampheol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1r,2r,3s,5r
6553875
(1R,3S,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol
CC1(C)C2CC1C(C)(O)C(O)C2
This item is not returnable. View return policy
22422-34-0
C10H18O2
170.25
MFCD09955216
MOILFCKRQFQVFS-UHFFFAOYNA-N
1r,2r,3s,5r---2,3-pinanediol, 1r,2r,3s,5r---pinanediol, 1r,2r,3s,5r-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, --2,3-pinanediol, unii-r58l0w3a75, --cis-pinane-cis-2,3-diol, pinanediol, --2-hydroxyisopinocampheol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1r,2r,3s,5r
6553875
(1R,3S,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol
CC1(C)C2CC1C(C)(O)C(O)C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 55.0°C to 60.0°C
Color Colorless to Yellow
Boiling Point 101.0°C to 102.0°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 06,4145
Specific Rotation Condition −8.50 (20.00°C c=6.5,toluene)
Specific Rotation − 8.50
Formula Weight 170.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (1R,2R,3S,5R)-(-)-2,3-Pinanediol
Show More Show Less
compliance-icons
Product Identifier
  • (1R,2R,3S,5R)-(-)-2,3-Pinanediol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'