missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-2-PCCA hydrochloride, MedChemExpress

Product Code. 16469436
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16469436

Brand: Medchem Express HY100013A1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (1R,2R)-2-PCCA hydrochloride is a diastereomer of 2-PCCA, and acts as a potent GPR88 receptor agonist, with an EC50 of 3 nM in cell-free assay, and 603 nM in cell assay.

(1R,2R)-2-PCCA hydrochloride is a diastereomer of 2-PCCA, and acts as a potent GPR88 receptor agonist, with an EC50 of 3 nM in cell-free assay, and 603 nM in cell assay.

Specifications

Chemical Name or Material (1R,2R)-2-PCCA hydrochloride
Physical Form Solid
CAS 1609563-71-4
Color Deep Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C30H39Cl2N3O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 50 mg/mL (94.60 mM; Need ultrasonic)
SMILES CCCC1=CC=C(C2=CC=C(N(C[C@@H](N)[C@@H](C)CC)C([C@H]3[C@H](C4=CC=CC=N4)C3)=O)C=C2)C=C1.[H]Cl.[H]Cl
Molecular Weight (g/mol) 528.56
Formula Weight 528.56
Percent Purity 98.08%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.