missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%

Product Code. 10243661 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10243661 5 g Glass bottle 5g
10671632 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10243661 Supplier Thermo Scientific Acros Supplier No. 334790050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 716-61-0 | C9H13N2O4 | 213.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 716-61-0
Molecular Formula C9H13N2O4
Molecular Weight (g/mol) 213.21
MDL Number MFCD00078126
InChI Key OCYJXSUPZMNXEN-RKDXNWHRSA-O
Synonym 1r,2r-2-amino-1-4-nitrophenyl propane-1,3-diol, chloramphenicol base, levoamine, 1r,2r---2-amino-1-4-nitrophenyl-1,3-propanediol, d---threo-2-amino-1-4-nitrophenyl-1,3-propanediol, d-threo---2-amino-1-p-nitrophenyl-1,3-propanediol, 1r,2r-2-amino-1-4-nitrophenyl-1,3-propanediol, d--threo-2-amino-1-p-nitrophenyl propane-1,3-diol, 1,3-propanediol, 2-amino-1-4-nitrophenyl-, 1r,2r, 1,3-propanediol, 2-amino-1-p-nitrophenyl-, d-threo--
PubChem CID 69745
ChEBI CHEBI:78660
IUPAC Name (1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-aminium
SMILES [NH3+][C@H](CO)[C@H](O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 160°C to 166°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 5 g
Specific Rotation -31
Formula Weight 212.2
Percent Purity 96%
Physical Form Powder
Chemical Name or Material (1R, 2R)-2-Amino-1-(4-nitrophenyl)propane-1, 3-diol, 96%
Show More Show Less
compliance-icons
Product Identifier
  • (1R,2R)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.