missing translation for 'onlineSavingsMsg'
Learn More

(1R)-(-)-10-Camphorsulfonic Acid, TRC

Product Code. 30338242
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30338242 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338242 Supplier Toronto Research Chemicals Supplier No. TRCC1750451G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1R)-(-)-10-Camphorsulfonic Acid
CAS 35963-20-3
Molecular Formula C10 H16 O4 S
Quantity 1 g
Synonym (-)-10-Camphorsulfonic Acid, Bicyclo[2.2.1]heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, (1R,4S)-, Bicyclo[2.2.1]heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, (1R)-, ((1R,4S)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonic acid, (-)-(1R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptane-1-methanesulfonic acid, (-)-10-Camphorsulfonic acid, (-)-Camphorsulfonic acid, (1R)-(-)-10-Camphorsulfonic acid, (1R)-10-Camphorsulfonic acid, (1R)-Camphorsulfonic acid, (R)-(-)-10-Camphorsulfonic acid, (R)-(-)-CSA, (R)-Camphor-10-sulfonic acid, L(-)-Camphor-10-sulfonic acid, l-10-Camphorsulfonic acid, l-Camphorsulfonic acid, Bicyclo[2.2.1]heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, (1R,4S)-, Bicyclo[2.2.1]heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, (1R)-, ((1R,4S)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonic acid, (-)-(1R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptane-1-methanesulfonic acid, (-)-10-Camphorsulfonic acid, (-)-Camphorsulfonic acid, (1R)-(-)-10-Camphorsulfonic acid, (1R)-10-Camphorsulfonic acid, (1R)-Camphorsulfonic acid, (R)-(-)-10-Camphorsulfonic acid, (R)-(-)-CSA, (R)-Camphor-10-sulfonic acid, L(-)-Camphor-10-sulfonic acid, l-10-Camphorsulfonic acid, l-Camphorsulfonic acid
InChI Formula InChI=1S/C10H16O4S/c1-9(2)7-3-4-10(9,8(11)5-7)6-15(12,13)14/h7H,3-6H2,1-2H3,(H,12,13,14)/t7-,10-/m0/s1
SMILES CC1(C)[C@H]2CC[C@]1(CS(=O)(=O)O)C(=O)C2
IUPAC Name [(1S,4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
Molecular Weight (g/mol) 232.3
Formula Weight 232.0769
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.