missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tool Steel, CPM 15V, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Tool Steel, CPM 15V, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Aluminium/Niobium/Arsenic/Antimony/Lead/Zirconium/Titanium/Tin/Sulfur/Cobalt/Phosphorus/Tungsten/Copper/Nickel/Manganese/Silicon/Molybdenum/Carbon/Chromium/Vanadium/Nitrogen/Oxygen/Iron |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Aluminium -200 ppm/Niobium -60 ppm/Arsenic -30 ppm/Antimony -20 ppm/Lead -20 ppm/Zirconium -20 ppm/Titanium 0.0027 %/Tin 0.0054 %/Sulfur 0.01 %/Cobalt 0.018 %/Phosphorus 0.022 %/Tungsten 0.08 ppm/Copper 0.084 %/Nickel 0.123 %/Manganese 0.39 %/Silicon 0.89 %/Molybdenum 1.24 %/Carbon 3.51 %/Chromium 5.5 %/Vanadium 15.6 %/Nitrogen (<1160) ppm/Oxygen (<150) ppm/Iron (72) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction