missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium, Ti 6-7, R56700, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Titanium, Ti 6-7, R56700, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Lead/Antimony/Hafnium/Magnesium/Palladium/Phosphorus/Boron/Tungsten/Ruthenium/Copper/Manganese/Zirconium/Hydrogen/Cobalt/Tantalum/Titanium/Molybdenum/Yttrium/Tin/Nitrogen/Chromium/Carbon/Nickel/Silicon/Vanadium/Iron/Oxygen/Aluminium/Niobium |
| CAS | 7440-32-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Lead (<0.0001) %/Antimony (<0.001) %/Hafnium (<0.001) %/Magnesium (<0.001) %/Palladium (<0.001) %/Phosphorus (<0.001) %/Boron (<0.002) %/Tungsten (<0.002) %/Ruthenium (<0.01) %/Copper (<0.02) %/Manganese (<0.02) %/Zirconium (<0.02) %/Hydrogen [(0.001)] %/Cobalt [(0.004)] %/Tantalum [(0.006)] %/Titanium [(86.5)] %/Molybdenum [0.0028] %/Yttrium [0.003] %/Tin [0.005] %/Nitrogen [0.0059] %/Chromium [0.007] %/Carbon [0.008] %/Nickel [0.011] %/Silicon [0.017] %/Vanadium [0.03] %/Iron [0.17] %/Oxygen [0.18] %/Aluminium [6.16] %/Niobium [6.85] % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction