missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium, Ti 3-2.5, Grade 9, R56320, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Titanium, Ti 3-2.5, Grade 9, R56320, Ø32x3mm
Specifications
Specifications
| Chemical Name or Material | Boron/Zirconium/Copper/Molybdenum/Nitrogen/Tin/Silicon/Carbon/Chromium/Nickel/Oxygen/Iron/Vanadium/Aluminium/Yttrium/Hydrogen/Ruthenium/Niobium/Manganese/Tungsten/Phosphorus/Titanium/Cobalt |
| CAS | 7440-32-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0004 %/Zirconium 0.002 %/Copper 0.003 %/Molybdenum 0.004 %/Nitrogen 0.004 %/Tin 0.004 %/Silicon 0.008 %/Carbon 0.011 %/Chromium 0.016 %/Nickel 0.023 %/Oxygen 0.083 %/Iron 0.19 %/Vanadium 2.23 %/Aluminium 2.98 %/Yttrium (0.0004) %/Hydrogen (0.001) %/Ruthenium (0.001) %/Niobium (0.002) %/Manganese (0.003) %/Tungsten (0.003) %/Phosphorus (0.004) %/Titanium (94.4) %/Cobalt <0.004 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction