missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, A485 Grade 3, K19965, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Alloy Steel, A485 Grade 3, K19965, Ø40x20mm
Specifications
Specifications
| Chemical Name or Material | Zinc/Tin/Nitrogen/Vanadium/Phosphorus/Aluminium/Copper/Nickel/Molybdenum/Silicon/Manganese/Carbon/Chromium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Zinc 0.0005 %/Tin 0.007 %/Nitrogen 0.008 %/Vanadium 0.009 %/Phosphorus 0.02 %/Aluminium 0.024 %/Copper 0.14 %/Nickel 0.14 %/Molybdenum 0.21 %/Silicon 0.247 %/Manganese 0.59 %/Carbon 0.93 %/Chromium 1.7 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction