missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, 1.8523, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Alloy Steel, 1.8523, Ø38x20mm
Specifications
Specifications
| Chemical Name or Material | Arsenic/Tin/Phosphorus/Nitrogen/Cobalt/Aluminium/Copper/Nickel/Vanadium/Silicon/Carbon/Manganese/Molybdenum/Chromium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Arsenic 0.004 %/Tin 0.0049 %/Phosphorus 0.006 %/Nitrogen 0.0155 %/Cobalt 0.0197 %/Aluminium 0.0208 %/Copper 0.0978 %/Nickel 0.1062 %/Vanadium 0.198 %/Silicon 0.25 %/Carbon 0.401 %/Manganese 0.604 %/Molybdenum 0.946 %/Chromium 3.29 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction