missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, AISI 1541, G15410, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Carbon Steel, AISI 1541, G15410, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Magnesium/Zirconium/Boron/Niobium/Calcium/Titanium/Aluminium/Antimony/Lead/Oxygen/Zinc/Tungsten/Arsenic/Cobalt/Nitrogen/Phosphorus/Tin/Vanadium/Molybdenum/Nickel/Chromium/Silicon/Copper/Carbon/Manganese |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Magnesium (<0.001) %/Zirconium (<0.002) %/Boron (0.0003) %/Niobium (0.001) %/Calcium (0.0012) %/Titanium (0.0015) %/Aluminium (0.002) %/Antimony (0.002) %/Lead (0.002) %/Oxygen (0.002) %/Zinc (0.003) %/Tungsten (0.004) %/Arsenic (0.005) %/Cobalt (0.008) %/Nitrogen (0.01) %/Phosphorus (0.011) %/Tin (0.015) %/Vanadium (0.027) %/Molybdenum (0.059) %/Nickel (0.178) %/Chromium (0.19) %/Silicon (0.19) %/Copper (0.3) %/Carbon (0.41) %/Manganese (1.48) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction