missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Aluminum Bronze D, CDA 614, C61400, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Aluminum Bronze D, CDA 614, C61400, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Phosphorus/Silver/Magnesium/Silicon/Nickel/Zinc/Manganese/Tin/Iron/Aluminium/Copper/Antimony/Lead |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Phosphorus 0.0008 %/Silver 0.001 %/Magnesium 0.005 %/Silicon 0.0124 %/Nickel 0.0242 %/Zinc 0.06 %/Manganese 0.082 %/Tin 0.301 %/Iron 2.74 %/Aluminium 6.81 %/Copper 89.99 %/Antimony (0.0004) %/Lead (0.0007) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction