missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Reinforcing Steel, A706 Grade 60, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Alloy Steel, Reinforcing Steel, A706 Grade 60, Ø32x19mm
Specifications
Specifications
| Chemical Name or Material | Titanium/Niobium/Aluminium/Tungsten/Arsenic/Nitrogen/Tin/Cobalt/Phosphorus/Vanadium/Molybdenum/Chromium/Nickel/Silicon/Carbon/Copper/Manganese/Bismuth/Cadmium/Magnesium/Selenium/Tantalum/Zirconium/Zinc/Oxygen/Calcium/Boron/Lead/Antimony |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Titanium 0.0011 %/Niobium 0.0016 %/Aluminium 0.0021 %/Tungsten 0.003 %/Arsenic 0.0058 %/Nitrogen 0.013 %/Tin 0.0132 %/Cobalt 0.014 %/Phosphorus 0.016 %/Vanadium 0.049 %/Molybdenum 0.055 %/Chromium 0.153 %/Nickel 0.182 %/Silicon 0.238 %/Carbon 0.243 %/Copper 0.307 %/Manganese 1.27 %/Bismuth (<0.0010) %/Cadmium (<0.0010) %/Magnesium (<0.0010) %/Selenium (<0.0010) %/Tantalum (<0.0010) %/Zirconium (<0.0020) %/Zinc (<0.0040) %/Oxygen (<0.0140) %/Calcium (0.0004) %/Boron (0.0005) %/Lead (0.002) %/Antimony (0.003) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction