missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Ni/Cr/Mo, AISI 8740, G87400, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Alloy Steel, Ni/Cr/Mo, AISI 8740, G87400, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Titanium/Niobium/Tungsten/Vanadium/Nitrogen/Arsenic/Tin/Cobalt/Phosphorus/Aluminium/Copper/Molybdenum/Silicon/Carbon/Nickel/Chromium/Manganese/Cadmium/Calcium/Zinc/Magnesium/Selenium/Tantalum/Bismuth/Boron/Antimony/Lead/Oxygen/Zirconium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Titanium 0.001 %/Niobium 0.0016 %/Tungsten 0.003 %/Vanadium 0.003 %/Nitrogen 0.0059 %/Arsenic 0.006 %/Tin 0.008 %/Cobalt 0.0086 %/Phosphorus 0.011 %/Aluminium 0.026 %/Copper 0.182 %/Molybdenum 0.21 %/Silicon 0.247 %/Carbon 0.406 %/Nickel 0.412 %/Chromium 0.463 %/Manganese 0.88 %/Cadmium (<0.001) %/Calcium (<0.001) %/Zinc (<0.002) %/Magnesium (<0.005) %/Selenium (<0.005) %/Tantalum (<0.008) %/Bismuth (<0.02) %/Boron (0.001) %/Antimony (0.003) %/Lead (0.003) %/Oxygen (0.003) %/Zirconium (0.003) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction