missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Ferritic Cr/Mo/V Steel, K91560, A182 Grade F91, K91560, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Alloy Steel, Ferritic Cr/Mo/V Steel, K91560, A182 Grade F91, K91560, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Titanium/Oxygen/Tungsten/Tin/Aluminium/Cobalt/Phosphorus/Copper/Nitrogen/Niobium/Carbon/Nickel/Vanadium/Silicon/Manganese/Molybdenum/Chromium/Antimony/Hafnium/Hydrogen/Lead/Magnesium/Yttrium/Zinc/Zirconium/Arsenic/Boron/Calcium/Palladium/Selenium/Tantalum |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Titanium 0.0021 %/Oxygen 0.003 %/Tungsten 0.003 %/Tin 0.004 %/Aluminium 0.006 %/Cobalt 0.013 %/Phosphorus 0.015 %/Copper 0.041 %/Nitrogen 0.046 %/Niobium 0.07 %/Carbon 0.099 %/Nickel 0.187 %/Vanadium 0.198 %/Silicon 0.27 %/Manganese 0.453 %/Molybdenum 0.94 %/Chromium 8.24 %/Antimony (<10) %/Hafnium (<10) %/Hydrogen (<10) %/Lead (<10) %/Magnesium (<10) %/Yttrium (<10) %/Zinc (<10) %/Zirconium (<40) %/Arsenic (<50) %/Boron (<50) %/Calcium (<50) %/Palladium (<50) %/Selenium (<50) %/Tantalum (<70) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction