missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, SUS, Al/Si Alloy, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Aluminum, SUS, Al/Si Alloy, Ø40x18mm
Specifications
Specifications
| Chemical Name or Material | Lead/Beryllium/Titanium/Cobalt/Zirconium/Antimony/Chromium/Strontium/Nickel/Tin/Iron/Zinc/Manganese/Copper/Magnesium/Silver/Silicon/Calcium/Lithium/Sodium/Phosphorus |
| CAS | 7429-90-5 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Lead 0.001 %/Beryllium 0.015 %/Titanium 0.02 %/Cobalt 0.025 %/Zirconium 0.025 %/Antimony 0.03 %/Chromium 0.06 %/Strontium 0.07 %/Nickel 0.12 %/Tin 0.13 %/Iron 0.25 %/Zinc 0.25 %/Manganese 0.45 %/Copper 0.75 %/Magnesium 0.9 %/Silver 1 %/Silicon 9.5 %/Calcium <0.001 %/Lithium <0.001 %/Sodium <0.001 %/Phosphorus <0.01 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction